Skip to Content
Merck
All Photos(3)

Key Documents

P22400

Sigma-Aldrich

N-Phenyldiethanolamine

97%

Synonym(s):

2,2′-(Phenylimino)diethanol, N,N-Bis(2-hydroxyethyl)aniline, N-Phenyl-2,2′-iminodiethanol

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$45.90
1 KG
$88.50

$45.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$45.90
1 KG
$88.50

About This Item

Linear Formula:
C6H5N(CH2CH2OH)2
CAS Number:
Molecular Weight:
181.23
Beilstein:
2096832
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$45.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

>1 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

97%

form

solid

mp

56-58 °C (lit.)

SMILES string

OCCN(CCO)c1ccccc1

InChI

1S/C10H15NO2/c12-8-6-11(7-9-13)10-4-2-1-3-5-10/h1-5,12-13H,6-9H2

InChI key

OJPDDQSCZGTACX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Król et al.
Colloid and polymer science, 289(15-16), 1757-1767 (2011-12-02)
WAXS, DSC and AFM methods were employed to compare phase structures of the coatings obtained from waterborne polyurethane cationomers which had been synthesised in the reaction of some diisocyanates (MDI, IPDI, TDI and HDI) with polyoxyethylene glycols (M = 600 and 2,000)
Aswin Anand Pai et al.
Clinical lymphoma, myeloma & leukemia, 20(2), 130-135 (2019-12-04)
High-dose melphalan (MEL) is the standard conditioning regimen used for autologous stem cell transplantation (ASCT) in patients with multiple myeloma (MM). Generic MEL is routinely used in various transplant centers across the world including ours due to its reduced cost

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service