Skip to Content
Merck
All Photos(1)

Key Documents

OCNQAK525

Sigma-Aldrich

SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots

fluorescence λem 525 nm, for 4 conjugation

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$750.00

$750.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$750.00

About This Item

UNSPSC Code:
43211915
NACRES:
NA.23

$750.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

fluorescence

λem 525 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Components:
  • QSA
  • Sulfo-SMCC
  • Traut′s Reagent
  • Coupling Buffer
  • Storage Buffer
  • Quenching Buffer
  • Tubes 4 x 1.7 mL

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service