Skip to Content
Merck
All Photos(1)

Key Documents

NCXAGPD50

Sigma-Aldrich

Silver Nanospheres

50 nm, NanoXact, Dried, PVP 40 kDa (Polymer), powder

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$175.00
5 MG
$653.00
10 MG
$1,100.00

$175.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$175.00
5 MG
$653.00
10 MG
$1,100.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

$175.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

nanospheres
powder

packaging

pkg of SKU size by silver mass

particle size

50 nm

Looking for similar products? Visit Product Comparison Guide

Application

Owing to their plasmonic properties nanoparticles of gold and silver find broad range of applications extending from diagnostics to molecular detection, photothermal and microwave-based therapies, and nanotoxicology. Colloidal silver nanoparticles also find applications in electronics and photovoltaics, and surface enhanced Raman spectroscopy (SERS) whereas the colloidal gold nanoparticles find applications in lateral flow assays (immunochromatographic assays) and as TEM labels and size standards.

Our NanoXact materials are precisely engineered, unagglomerated silver nanospheres with a narrow size distributions (CVs <15% for 30-200 nm sizes, <20% for 20 nm, and <25% for 5 and 10 nm sizes). These nanospheres contain PVP (40 kD polyvinylpyrrolidone) as excipient. The PVP enhances stability and dispersibility of NanpXact silver nanospheres. These are redispersible in a variety of polar solvents such as water, alcohols, DMF, and DMSO. They show medium dispersibility in chloroform, and low dispersibility in dichloromethane.

Storage and Stability

DO NOT FREEZE. Store at 2-25C in a dry place. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Legal Information

Product of nanoComposix, Inc.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Research on noble bimetallic nanoparticles for glucose biosensor detection.

Questions

  1. Does this material contain any organic substance on the surface of silver nanoparticles? Please explain.

    1 answer
    1. The particle surface comprises polyvinylpyrrolidone with a molecular weight of 40 kDa. The polyvinylpyrrolidone concentration is 90% of the particles by mass, and the silver concentration is 10% by mass.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service