Skip to Content
Merck
All Photos(1)

Key Documents

N26903

Sigma-Aldrich

2-Nitroterephthalic acid

≥99%

Synonym(s):

2-Nitro-1,4-benzenedicarboxylic acid, 2-Nitro-p-phthalic acid, 2-Nitrobenzene-1,4-dicarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$189.00

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$189.00

About This Item

Linear Formula:
O2NC6H3-1,4-(CO2H)2
CAS Number:
Molecular Weight:
211.13
Beilstein:
1913836
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99%

form

crystals

mp

270-272 °C (lit.)

SMILES string

OC(=O)c1ccc(C(O)=O)c(c1)[N+]([O-])=O

InChI

1S/C8H5NO6/c10-7(11)4-1-2-5(8(12)13)6(3-4)9(14)15/h1-3H,(H,10,11)(H,12,13)

InChI key

QUMITRDILMWWBC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Nurhasan et al.
Prostaglandins, leukotrienes, and essential fatty acids, 96, 57-61 (2015-03-11)
Fatty acid analysis requires standardized collection and storage of samples, which can be a challenge under field conditions. This study describes the effect of storage temperature on fatty acid composition in two sets of whole blood samples collected from 66
Rubén Angeriz-Campoy et al.
Bioresource technology, 194, 291-296 (2015-07-27)
Bio-hydrogen production from dry thermophilic anaerobic co-digestion (55°C and 20% total solids) of organic fraction of municipal solid waste (OFMSW) and food waste (FW) was studied. OFMSW coming from mechanical-biological treatment plants (MBT plants) presents a low organic matter concentration.
Adam H Metherel et al.
Lipids, 50(9), 895-905 (2015-08-04)
Dried blood spots (DBS) by fingertip prick collection for fatty acid profiling are becoming increasingly popular due to ease of collection, minimal invasiveness and its amenability to high-throughput analyses. Herein, we assess a microwave-assisted direct transesterification method for the production
Gina L Lonati et al.
The Journal of experimental biology, 218(Pt 16), 2620-2630 (2015-08-21)
Understanding toothed whale (odontocete) diving gas dynamics is important given the recent atypical mass strandings of odontocetes (particularly beaked whales) associated with mid-frequency naval sonar. Some stranded whales have exhibited gas emboli (pathologies resembling decompression sickness) in their specialized intramandibular
Ryan M Bradley et al.
Biochimica et biophysica acta, 1851(12), 1566-1576 (2015-09-30)
The acylglycerophosphate acyltransferase/lysophosphatidic acid acyltransferase (AGPAT/LPAAT) family is a group of homologous acyl-CoA-dependent lysophospholipid acyltransferases. We performed studies to better understand the subcellular localization, activity, and in vivo function of AGPAT4/LPAATδ, which we found is expressed in multiple mouse brain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service