Skip to Content
Merck
All Photos(2)

Key Documents

N1909

Sigma-Aldrich

1-Naphthoic acid

96%

Synonym(s):

1-Naphthalenecarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$52.50

$52.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$52.50

About This Item

Linear Formula:
C10H7CO2H
CAS Number:
Molecular Weight:
172.18
Beilstein:
1908896
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$52.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

96%

form

powder

bp

300 °C (lit.)

mp

157-160 °C (lit.)

SMILES string

OC(=O)c1cccc2ccccc12

InChI

1S/C11H8O2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,(H,12,13)

InChI key

LNETULKMXZVUST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Naphthoic acid can be used as a reactant to prepare:
  • Perinaphthenones by dehydrative annulation with alkynes in the presence of rhodium catalyst.[1]
  • Isocoumarin derivatives by reacting with 2-butyne via aerobic oxidative cyclization using Rh catalyst.[2]
  • N-Methoxy-N-methyl-1-naphthalenecarboxamide (Weinreb amide) by reacting with N,O-dimethylhydroxylamine and phosphorus trichloride.[3]
  • 1,4-Dihydro-1-naphthalenecarboxylic acid by Birch reduction.[4]

Other Notes

Remainder 2-naphthoic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Chandra et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(3), 704-713 (2009-09-02)
The Fourier transform infrared gas phase spectrum of Naphthoic acid (NA) was recorded in the region 4000-400 cm(-1). The Fourier transform Raman spectrum and Fourier transform IR spectra of NA were recorded in solid phase. Quantum chemical calculations of energies
Hiromasa Uchiyama et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 43(1-2), 71-77 (2011-04-06)
Spray-dried particles (SDPs) with indomethacin (IND) and alpha-glycosyl transferase-treated stevia (Stevia-G) indicated extremely high dissolution rates and apparent solubility compared to particles of a ground mixture and a physical mixture of IND/Stevia-G. The apparent solubility of IND from SDPs was
Synthesis of perinaphthenones through rhodium-catalyzed dehydrative annulation of 1-naphthoic acids with alkynes
Fukuyama T, et al.
Organic & Biomolecular Chemistry, 16, 7583-7587 (2018)
Qunfei Zhao et al.
Chemistry & biology, 15(7), 693-705 (2008-07-19)
Azinomycin B is a complex natural product containing densely assembled functionalities with potent antitumor activity. Cloning and sequence analysis of the azi gene cluster revealed an iterative type I polyketide synthase (PKS) gene, five nonribosomal peptide synthetases (NRPSs) genes and
Rajesh Sunasee et al.
The Journal of organic chemistry, 73(20), 8016-8020 (2008-09-26)
A method is described for converting tert-butyl benzoates or tert-butyl 1-naphthoates into derivatives having an alkyl or substituted alkyl group in a 1,4-relationship to an alkyl, aryl, alkenyl, or alkynyl group. Key steps in the sequence are (i) addition of

Questions

  1. How can it be used as plant root development

    1 answer
    1. Product N1909 is intended for use as a reagent in chemical synthesis rather than for use as a plant nutrient. Unfortunately, information is not available for the usage of this product for plant root development.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service