Skip to Content
Merck
All Photos(2)

Key Documents

N13054

Sigma-Aldrich

4-Nitrobenzyl bromide

99%

Synonym(s):

α-Bromo-4-nitrotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$61.60
100 G
$86.20

$61.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$61.60
100 G
$86.20

About This Item

Linear Formula:
O2NC6H4CH2Br
CAS Number:
Molecular Weight:
216.03
Beilstein:
742796
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$61.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

mp

96-99 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(CBr)cc1

InChI

1S/C7H6BrNO2/c8-5-6-1-3-7(4-2-6)9(10)11/h1-4H,5H2

InChI key

VOLRSQPSJGXRNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allergic contact dermatitis from para-nitrobenzyl bromide.
R Thompson et al.
Contact dermatitis, 38(4), 232-232 (1998-06-20)
Qing-Yuan Chen et al.
Di 1 jun yi da xue xue bao = Academic journal of the first medical college of PLA, 23(3), 273-276 (2003-03-26)
To synthesize p, p'-divinylazobenzene (DVAB). Wittig reagent was initially synthesized using P-nitrobenzyl bromide and triphenylphosphine, followed by reaction with formaldehyde to produce p-nitro styrene, which was then deoxidized by Zn/NaOH for the final product of DVAB. Fourier transform infrared spectroscopy
S Giorgini et al.
Italian general review of dermatology, 15(2), 107-112 (1978-05-01)
A case of occupational contact dermatitis in a female chemical pharmaceutical laboratory researcher is described. The allergological investigation revealed a state of sensitization to three mono-nitrobenzenes with halogen substitutions in the nucleus or a side chain. Problems related to sensitization
Ihor Kulai et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(63), 16066-16077 (2017-08-31)
Eight alkyl triarylstannanecarbodithioates were synthesized starting from the corresponding triarylstannyl chlorides. They were fully characterized by IR and

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service