MT51344712
Mettler-Toledo perfectION™ combination electrode
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
epoxy shaft
glass membrane (solid state)
description
BNC connector
Reference electrolyte Ion Electrolyte D
manufacturer/tradename
Mettler-Toledo 51344712
temperature measuring range
0-80 °C
cable L
1.2 m
pH range
2-12
Looking for similar products? Visit Product Comparison Guide
General description
Mettler Toledo perfectION™ ISE Combined ion-selective electrode means ISE measurement has never been so simple!
The new Click & Clear™ junction of the combined ion-selective electrodes combines three advantages in one tool: The electrolyte solution can flow easily through the junction by pressing the electrode head downwards. This ensures that the junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled if necessary.
Click & Clear™ is a sleeve junction that enables optimum contact between the electrolyte and measurement solution, even in difficult samples.
Features and Benefits
All combined ion-selective electrodes consist of a measuring electrode complete with an integrated reference. There is therefore no need for an additional reference electrode.
The new Click & Clear™ junction of the combined ion-selective electrodes combines three advantages in one tool: The electrolyte solution can flow easily through the junction by pressing the electrode head downwards. This ensures that the junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled if necessary.
Click & Clear™ is a sleeve junction that enables optimum contact between the electrolyte and measurement solution, even in difficult samples.
Features and Benefits
All combined ion-selective electrodes consist of a measuring electrode complete with an integrated reference. There is therefore no need for an additional reference electrode.
- Sensor with attached cable and BNC connector
- Reference electrolyte (60 mL)
- Operating instructions
To ensure an unproblematic start to your ISE measurement, the required calibration standards and ISA solutions (Ionic Strength Adjuster) can be ordered at the same time.
comb Cu
Legal Information
Click & Clear is a trademark of Mettler-Toledo, Inc.
perfectION is a trademark of Mettler-Toledo, Inc.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service