MT51344703
Mettler-Toledo perfectION™ combination electrode
About This Item
Recommended Products
material
epoxy shaft
glass membrane (polymer)
description
BNC connector
Reference electrolyte Ion Electrolyte A
manufacturer/tradename
Mettler-Toledo 51344703
temperature measuring range
0-40 °C
cable L
1.2 m
pH range
2.5-11
Looking for similar products? Visit Product Comparison Guide
General description
The new Click & Clear™ junction of the combined ion-selective electrodes combines three advantages in one tool: The electrolyte solution can flow easily through the junction by pressing the electrode head downwards. This ensures that the junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled if necessary.
Click & Clear™ is a sleeve junction that enables optimum contact between the electrolyte and measurement solution, even in difficult samples.
Features and Benefits
All combined ion-selective electrodes consist of a measuring electrode complete with an integrated reference. There is therefore no need for an additional reference electrode.
- Sensor with attached cable and BNC connector
- Reference electrolyte (60 mL)
- Operating instructions
To ensure an unproblematic start to your ISE measurement, the required calibration standards and ISA solutions (Ionic Strength Adjuster) can be ordered at the same time.
Legal Information
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service