Skip to Content
Merck
All Photos(1)

Key Documents

MT51343190

Mettler-Toledo InLab® Reference Routine electrode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
39121436
NACRES:
NB.79
Pricing and availability is not currently available.

material

glass shaft

description

Reference electrolyte 3 mol/L KCl
Reference system ARGENTHAL with Ag+ -trap
S7 connector

manufacturer/tradename

Mettler-Toledo 51343190

temperature measuring range

0-100 °C

L × diam.

120 mm × 12 mm

General description

InLab® Reference is a glass-body, reference electrode with S7 screw head. Filled with 3 mol/L KCl for routine applications.

Silver ion trap
The electrolyte in conventional silver/silver chloride reference systems must be saturated with silver chloride. With critical media such as those containing sulphides or proteins, this will result in diaphragm contamination with silver sulphide.

ARGENTHAL reference
This cannot happen with InLab electrodes which have an ARGENTHAL lead off and silver ion trap that ensure the electrolyte remains completely free from silver ions. The ARGENTHAL reference system also improves service life and measurement accuracy at differing sample temperatures.

Glass shaft
The glass shaft provides resistance against organic and corrosive samples.

Detachable cable
Different cables can be connected to the S7 screw head allowing use with a wide range of meters.

Legal Information

ARGENTHAL is a trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service