Skip to Content
Merck
All Photos(1)

Key Documents

M49887

Sigma-Aldrich

1-Methylhydantoin

97%

Synonym(s):

1-Methylimidazolidine-2,4-dione, Dioxy-creatinine, NSC 80560

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$141.00

$141.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$141.00

About This Item

Empirical Formula (Hill Notation):
C4H6N2O2
CAS Number:
Molecular Weight:
114.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$141.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

156-157 °C (lit.)

SMILES string

CN1CC(=O)NC1=O

InChI

1S/C4H6N2O2/c1-6-2-3(7)5-4(6)8/h2H2,1H3,(H,5,7,8)

InChI key

RHYBFKMFHLPQPH-UHFFFAOYSA-N

Application

Reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide

Reactant for synthesis of:
  • Selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition
  • Allosteric glucokinase activators
  • Hydantoin derivatives with antiproliferative activity
  • Thiohydantoins
  • P2X7 receptor antagonists

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Ienaga et al.
Biochimica et biophysica acta, 967(3), 441-443 (1988-12-15)
The metabolic pathway of 1-methylhydantoin (2) via 5-hydroxy-1-methylhydantoin (3), methylparabanic acid (4) and N5-methyloxaluric acid (5) proved to be a major and general one in mammals. Hence the formation of (3), which has not been detected in normal tissue, is
[1-Methylhydantoin, an unexpected metabolite of the intelligence-affecting substance dupracetam (author's transl)].
H D Dell et al.
Archiv der Pharmazie, 314(8), 697-702 (1981-08-01)
Hybrid biosensor for clinical and fermentation process control.
I Karube et al.
Annals of the New York Academy of Sciences, 434, 508-511 (1984-01-01)
P Fossati et al.
Clinical chemistry, 40(1), 130-137 (1994-01-01)
We describe an improved enzymatic ultraviolet absorbance method for assaying creatinine in serum, plasma, and urine. Creatinine is hydrolyzed by creatinine iminohydrolase (EC 3.5.4.21) to ammonia and N-methylhydantoin. The ammonia produced combines with 2-oxoglutarate and NADPH in the presence of
Bo Yang et al.
Renal failure, 29(8), 1025-1029 (2007-12-11)
1-Methylhydantoin is produced by bacterial creatinine deaminase in the intestinal tract of uremic patients and retaken up into the body. The present study was designed to explore the toxic effect of 1-methylhydantoin on renal proximal tubular cells in vitro. HK-2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service