Skip to Content
Merck
All Photos(2)

Key Documents

M47353

Sigma-Aldrich

mono-Methyl glutarate

95%

Synonym(s):

Monomethyl glutarate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$69.70
25 G
$331.00

$69.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$69.70
25 G
$331.00

About This Item

Linear Formula:
HOOC(CH2)3COOCH3
CAS Number:
Molecular Weight:
146.14
Beilstein:
1762381
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$69.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.438 (lit.)

bp

150-151 °C/10 mmHg (lit.)

density

1.169 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCCC(O)=O

InChI

1S/C6H10O4/c1-10-6(9)4-2-3-5(7)8/h2-4H2,1H3,(H,7,8)

InChI key

IBMRTYCHDPMBFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Abrupt changes in the concentrations of five prostatic tumor-associated markers in serum after acute myocardial infarction.
D Hardeman et al.
Clinical chemistry, 38(4), 605-607 (1992-04-01)
Issan Zhang et al.
Cell death discovery, 4, 41-41 (2018-10-12)
Glioblastoma multiforme is one of the most aggressive brain tumors and current therapies with temozolomide or suberoylanilide hydroxamic acid (SAHA, vorinostat) show considerable limitations. SAHA is a histone deacetylase (HDAC) inhibitor that can cause undesirable side effects due to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service