Skip to Content
Merck
All Photos(1)

Key Documents

M4031

Sigma-Aldrich

p-Menthane-1,8-diol monohydrate

Synonym(s):

cis-4-Hydroxy-α,α,4-trimethylcyclohexanemethanol monohydrate, cis-Terpin monohydrate, cis-p-Menthane-1,8-diol monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O2 · H2O
CAS Number:
Molecular Weight:
190.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

SMILES string

O.CC(C)(O)[C@@H]1CC[C@](C)(O)CC1

InChI

1S/C10H20O2.H2O/c1-9(2,11)8-4-6-10(3,12)7-5-8;/h8,11-12H,4-7H2,1-3H3;1H2/t8-,10+;

InChI key

JGKJMBOJWVAMIJ-OFAZAQPOSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fredros O Okumu et al.
Malaria journal, 8, 150-150 (2009-07-09)
Gold-standard tests of mosquito repellents involve exposing human volunteers to host-seeking mosquitoes, to assess the protective efficacy of the repellents. These techniques are not exposure-free and cannot be performed prior to toxicological evaluation. It is postulated that synthetic lures could
Thomas G T Jaenson et al.
Journal of medical entomology, 43(4), 731-736 (2006-08-09)
MyggA Natural (Bioglan, Lund, Sweden) is a commercially available repellent against blood-feeding arthropods. It contains 30% of lemon-scented eucalyptus, Corymbia citriodora (Hook.) K. D. Hill & L. A. S. Johnson (Myrtaceae), oil with a minimum of 50% p-menthane-3,8-diol. MyggA Natural
TERPIN hydrate.
Bulletin. American Pharmaceutical Association. Committee on National Formulary, 14, 41-41 (1946-03-01)
J K Trigg
Journal of the American Mosquito Control Association, 12(2 Pt 1), 243-246 (1996-06-01)
A eucalyptus-based insect repellent (PMD) with the principal active ingredient p-menthane-3,8-diol was evaluated in the field in comparison with deet. In human landing catches in Tanzania, 3 formulations of PMD were tested against Anopheles gambiae and An. funestus. Repellents, applied
Samira S Garboui et al.
Experimental & applied acarology, 40(3-4), 271-277 (2006-11-15)
In the field in south-central Sweden, we tested by randomised, standardised methodology the potential anti-tick repellent activity of two concentrations of MyggA Natural spray (containing PMD) (4.2 and 3.2 g/m2) and one of RB86 (with 70% neem oil containing azadirachtin)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service