Skip to Content
Merck
All Photos(1)

Documents

M29401

Sigma-Aldrich

2-Methylanthracene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12
CAS Number:
Molecular Weight:
192.26
Beilstein:
2042447
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

mp

204-206 °C (lit.)

SMILES string

Cc1ccc2cc3ccccc3cc2c1

InChI

1S/C15H12/c1-11-6-7-14-9-12-4-2-3-5-13(12)10-15(14)8-11/h2-10H,1H3

InChI key

GYMFBYTZOGMSQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gareth E Thomas et al.
Frontiers in microbiology, 11, 1706-1706 (2020-08-09)
In September 2017 the Agia Zoni II sank in the Saronic Gulf, Greece, releasing approximately 500 tonnes of heavy fuel oil, contaminating the Salamina and Athens coastlines. Effects of the spill, and remediation efforts, on sediment microbial communities were quantified
Jolanta Kumirska et al.
Chemosphere, 232, 232-242 (2019-06-04)
In this paper, an analytical method for the simultaneous determination of twenty pharmaceuticals (eight non-steroidal anti-inflammatory drugs, five oestrogenic hormones, two antiepileptic drugs, two β-blockers, and three antidepressants) in soils was developed. The optimal method included ultrasound-assisted extraction, a clean-up
Mei-Hui Tai et al.
International journal of cancer, 120(9), 1855-1862 (2007-02-03)
Smoking is a well-documented risk factor for the development of pancreatic adenocarcinoma. Although the most abundant polycyclic aromatic hydrocarbons (PAHs) in cigarette smoke are methylated anthracenes and phenanthrenes, the epigenetic toxicity of these compounds has not been extensively studied. We
Samuel J Shin et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(52), 32939-32946 (2020-12-16)
The faradaic reaction at the insulator is counterintuitive. For this reason, electroorganic reactions at the dielectric layer have been scarcely investigated despite their interesting aspects and opportunities. In particular, the cathodic reaction at a silicon oxide surface under a negative
Capillary gas chromatography/pulsed supersonic jet/fluorescence excitation spectroscopy for the identification of methylanthracenes in a complex environmental sample.
B V Pepich et al.
Analytical chemistry, 58(13), 2825-2830 (1986-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service