Skip to Content
Merck
All Photos(1)

Key Documents

M1626

Sigma-Aldrich

6-Methoxyharmalan

crystalline

Synonym(s):

4,9-Dihydro-6-methoxy-1-methyl-3H-pyrido(3,4-b)indole

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$93.10

$93.10


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$93.10

About This Item

Empirical Formula (Hill Notation):
C13H14N2O
CAS Number:
Molecular Weight:
214.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$93.10


Estimated to ship onMay 29, 2025


Request a Bulk Order

form

crystalline

color

yellow

storage temp.

2-8°C

SMILES string

COc1ccc2[nH]c3C(C)=NCCc3c2c1

InChI

1S/C13H14N2O/c1-8-13-10(5-6-14-8)11-7-9(16-2)3-4-12(11)15-13/h3-4,7,15H,5-6H2,1-2H3

InChI key

HMBHRMFLDKKSCT-UHFFFAOYSA-N

Application

  • Reactant for preparation of TRPV1-dependent vasodilator

Biochem/physiol Actions

Weak serotonin inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V P Poshivalov
Biulleten' eksperimental'noi biologii i meditsiny, 99(3), 323-325 (1985-03-01)
The ethological spectrum of four beta-carbolines was studied for intraspecific aggression, sociability integration and individual behavior. It was established that beta-carboline-3-carboxyethyl ether given at subconvulsant doses increases aggression, lowers sociability as a diazepam antagonist. Diazepam reduces aggression and increases sociability.
Fernando D Villarruel et al.
Physical chemistry chemical physics : PCCP, 22(36), 20901-20913 (2020-09-12)
3,4-Dihydro-β-carbolines (DHβCs) are a set of endogenously synthesized alkaloids spread over a great variety of living species (e.g., plants, animals and microorganisms), playing a broad spectrum of biological, biochemical and/or pharmacological roles, in a structure-dependent manner. Addressing unresolved fundamental aspects
M Micaela Gonzalez et al.
International journal of antimicrobial agents, 52(4), 459-468 (2018-07-15)
Several commercial and novel synthetic β-carbolines (βCs) were evaluated for their antiviral activity against herpes simplex virus type 1 (HSV-1) using an adapted MTS assay. Of 21 drugs tested, although 11 exerted antiviral activity at non-cytotoxic concentrations, only 3 of
Sudeep Karki et al.
Analytical biochemistry, 587, 113463-113463 (2019-10-02)
Synaptic adhesion molecules, including presynaptic neurexins (NRXNs) and post-synaptic leucine-rich repeat transmembrane (LRRTM) proteins are important for development and maintenance of brain neuronal networks. NRXNs are probably the best characterized synaptic adhesion molecules, and one of the major presynaptic organizer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service