Skip to Content
Merck
All Photos(3)

Key Documents

M15451

Sigma-Aldrich

5-Methoxy-2-methylindole

5-Methoxy-2-methylindole

99%

Synonym(s):

NSC 63817

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$67.90
5 G
$225.00

$67.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$67.90
5 G
$225.00

About This Item

Empirical Formula (Hill Notation):
C10H11NO
CAS Number:
Molecular Weight:
161.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$67.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

mp

86-88 °C (lit.)

SMILES string

COc1ccc2[nH]c(C)cc2c1

InChI

1S/C10H11NO/c1-7-5-8-6-9(12-2)3-4-10(8)11-7/h3-6,11H,1-2H3

InChI key

VSWGLJOQFUMFOQ-UHFFFAOYSA-N

Application

reactant in preparation of indolylquinoxalines by condensation reactions
reactant in preparation of alkylindoles via Ir-catalyzed reductive alkylation
reactant in arylation reactions using a palladium acetate catalyst
reactant in enantioselective Friedel-Crafts alkylation
reactant in stereoselective synthesis of cyclopentaindolones via stereoselective [3+2] cyclopentannulation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca R Helm et al.
PloS one, 12(12), e0188601-e0188601 (2017-12-28)
Many animals go through one or more metamorphoses during their lives, however, the molecular underpinnings of metamorphosis across diverse species are not well understood. Medusozoa (Cnidaria) is a clade of animals with complex life cycles, these life cycles can include
Luna Yamamori et al.
Zoological science, 34(3), 173-178 (2017-06-08)
Scyphozoa, Cubozoa and Hydrozoa are classes in the phylum Cnidaria that undergo metagenesis involving a dramatic morphological transition. In Scyphozoa and Cubozoa, when exposed to species- or strain-specific transition-inducing stimuli, asexually reproducing benthic polyps transform into sexually reproducing planktonic medusae.

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service