Skip to Content
Merck
All Photos(1)

Key Documents

M13203

Sigma-Aldrich

3-Methoxycatechol

99%

Synonym(s):

1,2-Dihydroxy-3-methoxybenzene, 3-Methoxypyrocatechol, Pyrogallol monomethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$99.70

$99.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$99.70

About This Item

Linear Formula:
CH3OC6H3(OH)2
CAS Number:
Molecular Weight:
140.14
Beilstein:
1909165
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$99.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

bp

146-147 °C/15 mmHg (lit.)

mp

38-43 °C (lit.)

SMILES string

COc1cccc(O)c1O

InChI

1S/C7H8O3/c1-10-6-4-2-3-5(8)7(6)9/h2-4,8-9H,1H3

InChI key

LPYUENQFPVNPHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keith M Anderson et al.
The Journal of organic chemistry, 72(26), 9875-9880 (2007-11-30)
A novel universal support for deoxyribo- and ribonucleic acid synthesis has been developed. The support, constructed from 1,4-dimethoxycatechol, represents an improvement over existing universal supports because of its ability to cleave and deprotect under mild conditions in standard reagents. Because
M Hirose et al.
Japanese journal of cancer research : Gann, 81(9), 857-861 (1990-09-01)
The effects of sodium nitrite (NaNO2) and catechol or 3-methoxycatechol in combination were examined in male F344 rats. Animals were treated with 0.3% NaNO2 in the drinking water and 0.8% catechol or 2% 3-methoxycatechol in powdered diet for 24 weeks.
M Kawabe et al.
Japanese journal of cancer research : Gann, 85(1), 17-25 (1994-01-01)
The effects of combined treatment with NaNO2 and phenolic compounds on N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) stomach carcinogenesis were investigated in F344 rats. In the first experiment, groups of 15-20 male rats were treated with an intragastric dose of 150 mg/kg body weight
Huayun Deng et al.
Pharmaceuticals (Basel, Switzerland), 6(4), 500-509 (2013-11-28)
Nearly 1% of all clinically used drugs are catecholics, a family of catechol-containing compounds. Using label-free dynamic mass redistribution and Tango β-arrestin translocation assays, we show that several catecholics, including benserazide, catechol, 3-methoxycatechol, pyrogallol, (+)-taxifolin and fenoldopam, display agonistic activity
M Hirose et al.
Cancer research, 53(1), 32-37 (1993-01-01)
Effects of simultaneous treatment with NaNO2 and butylated hydroxyanisole, catechol, or 3-methoxycatechol were examined in a rat multiorgan carcinogenesis model. Groups of 15 animals were given a single i.p. injection of 100 mg/kg of body weight diethylnitrosamine, 4 i.p. injections

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service