Skip to Content
Merck
All Photos(1)

Key Documents

M0137

Sigma-Aldrich

Magnesium iodate tetrahydrate

99.0%

Synonym(s):

Bisiodic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(IO3)2 · 4H2O
CAS Number:
Molecular Weight:
446.17
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
powder

Assay

99.0%

form

powder

SMILES string

[Mg].OI(=O)=O

InChI

1S/2HIO3.Mg.4H2O/c2*2-1(3)4;;;;;/h2*(H,2,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

QRDFCLGYWATLRM-UHFFFAOYSA-L

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allen E Haddrell et al.
The journal of physical chemistry. A, 116(40), 9941-9953 (2012-09-18)
The most used instrument in single particle hygroscopic analysis over the past thirty years has been the electrodynamic balance (EDB). Two general assumptions are made in hygroscopic studies involving the EDB. First, it is assumed that the net charge on
Barbara Krassowska-Swiebocka et al.
Molecules (Basel, Switzerland), 10(2), 394-400 (2007-11-17)
Benzene, halobenzenes, and a number of more or less deactivated arenes,including nitrobenzene, readily reacted in anhydrous HIO3/AcOH/Ac2O/conc. H2SO4 mixtures to probably give ArIO2 intermediates or other hypervalent species (not isolated). The final reaction mixtures were poured into excess aq. Na2SO3
Amanda C Bean et al.
Inorganic chemistry, 44(15), 5209-5211 (2005-07-19)
The hydrothermal reaction of a (239)Pu(IV) stock solution in the presence of iodic acid and 1 M KOH produces reddish-brown single crystals of (PuO(2))(2)(IO(3))(OH)(3). The structure consists of two-dimensional layers forming in the ac plane and is the first single-crystal
Colour reactions of iodic acid as reagent for identifying drugs by thin-layer chromatography.
G Cavazzutti et al.
Journal of chromatography, 268(3), 528-534 (1983-10-07)
[Correlation between ERPF and retention function on 131I-OIH renogram].
M Maeda et al.
Kaku igaku. The Japanese journal of nuclear medicine, 21(10), 1285-1292 (1984-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service