Skip to Content
Merck
All Photos(1)

Key Documents

LGFR1000

Sigma-Aldrich

Red Fluorescent PLGA nanoparticles

1 μm average diameter

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$677.00

$677.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$677.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

$677.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

Excitation/Emission: 530/582 nm
Excitation/Emission: 460/500 nm

form

pellets

average diameter

1 μm

Looking for similar products? Visit Product Comparison Guide

General description

Particles made of poly(D,L-lactide-co-glycolide) L/G ratio of 50/50, inherent viscosity of ~0.45 dL/g.To re-constitute:
  • Warm to ambient temperature
  • Add DI water or buffer to achieve desired concentration
  • Vortex and sonicate for 5 minutes to ensure particles are well suspended

Application

Fluorescent PLGA particles for imaging and cell tracking in drug discovery. Particles can be used for fluorscent assays and drug delivery.

Legal Information

Product of Phosphorex, Inc.Degradex(RM) is a product of Phosphorex, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Questions

  1. Can this product be resuspended in water? What is its stability lifetime?

    1 answer
    1. LGFR100 is provided in lyophilized powder form and should be stored at -20°C in a dry environment protected from light. It can be resuspended in water by following these steps: remove the product from the freezer, wait until the temperature rises to ambient, add DI water or buffer to the container, vortex and sonicate for 5 minutes to ensure the microspheres are well suspended.

      It is strongly recommended to use the product immediately upon resuspension because the particles begin to degrade as soon as they are introduced into an aqueous solution. If storage of particles in suspension is required, it is recommended to store them at 2-8 °C to slow the degradation.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service