Skip to Content
Merck
All Photos(1)

Documents

K3603

Sigma-Aldrich

6,7-Dihydro-4-benzo[b]thiophenone

98%

Synonym(s):

4-Keto-4,5,6,7-tetrahydrothianaphthene, 4-Oxo-4,5,6,7-tetrahydrobenzo[b]thiophene, 4-Oxo-4,5,6,7-tetrahydrothianaphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8OS
CAS Number:
Molecular Weight:
152.21
Beilstein:
112276
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

mp

38-40 °C (lit.)

SMILES string

O=C1CCCc2sccc12

InChI

1S/C8H8OS/c9-7-2-1-3-8-6(7)4-5-10-8/h4-5H,1-3H2

InChI key

GJEKNELSXNSYAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service