Skip to Content
Merck
All Photos(1)

Key Documents

JKA0002

Sigma-Aldrich

Y-PEG40K

average MN 40,000, MAL, thiol reactive

Synonym(s):

Y-PEG-MAL

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$937.00

$937.00


Estimated to ship onApril 08, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$937.00

About This Item

Linear Formula:
(CH3O(CH2CH2O)nCH2CH2)(CH3O(CH2CH2O)nCH2CO)NCH2CONHCH2CH2NHCOCH2CH2C4H4O2N
UNSPSC Code:
12162002
NACRES:
NA.23

$937.00


Estimated to ship onApril 08, 2025


Request a Bulk Order

Product Name

Y-PEG40K-MAL, average Mn 40,000

form

solid

mol wt

average Mn 40,000

reaction suitability

reactivity: thiol reactive

polymer architecture

shape: Y-shape
functionality: monofunctional

storage temp.

−20°C

General description

Y-shape Maleimide PEG. Thiol reactive PEG ; reacts at pH 5.0-6.5. JenKem Technology′s patented products, Y-shape PEG derivatives, also known as branched PEG derivatives, contain two linear methoxy PEG chains attached to a central active core. JenKem proprietary Y-shape PEGs are more selective, due to their sterically bulky structure.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resch, G., P. Moreillon, and V.A. Fischetti
AMB Express, 1(29), 1-5 (2011)

Articles

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service