Skip to Content
Merck
All Photos(1)

Key Documents

I22005

Sigma-Aldrich

Isopropyl disulfide

96%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$84.70

$84.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$84.70

About This Item

Linear Formula:
[(CH3)2CH]2S2
CAS Number:
Molecular Weight:
150.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$84.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

96%

refractive index

n20/D 1.4906 (lit.)

bp

175-176 °C (lit.)

density

0.943 g/mL at 25 °C (lit.)

SMILES string

CC(C)SSC(C)C

InChI

1S/C6H14S2/c1-5(2)7-8-6(3)4/h5-6H,1-4H3

InChI key

LZAZXBXPKRULLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bofan Yu et al.
Journal of chromatography. A, 1356, 221-229 (2014-07-07)
For the expanded application area, fast trace analysis of certain high boiling point (i.e., 150-250 °C) volatile organic compounds (HVOCs) in water, a large volume-direct aqueous injection-gas chromatography (LV-DAI-GC) method was optimized for the following parameters: packed sorbent for sample

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service