Skip to Content
Merck
All Photos(1)

Key Documents

I10007

Sigma-Aldrich

3-Iodophenol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$53.80
25 G
$144.00

$53.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$53.80
25 G
$144.00

About This Item

Linear Formula:
IC6H4OH
CAS Number:
Molecular Weight:
220.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$53.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

42-44 °C (lit.)

storage temp.

2-8°C

SMILES string

Oc1cccc(I)c1

InChI

1S/C6H5IO/c7-5-2-1-3-6(8)4-5/h1-4,8H

InChI key

FXTKWBZFNQHAAO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Achintya Jana et al.
Dalton transactions (Cambridge, England : 2003), 46(6), 1986-1995 (2017-01-24)
The synthesis and characterization of a new pyrazine-based "flexible" and ditopic platinum(ii) organometallic molecule (3) is being reported. Flexibility in this molecule is due to the presence of ether functional groups that bridge the rigid core and periphery. Due to
Lihua Yang et al.
PloS one, 10(7), e0131193-e0131193 (2015-07-15)
A luminol-H2O2-HRP chemiluminescence system with high relative luminescent intensity (RLU) and long stabilization time was investigated. First, the comparative study on the enhancement effect of ten compounds as enhancers to the luminol-H2O2-HRP chemiluminescence system was carried out, and the results

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service