Skip to Content
Merck
All Photos(1)

Key Documents

HAM79433

Supelco

Hamilton® PRP-X100 (10 µm) HPLC Columns

L x I.D. 250 mm x 4.1 mm

Synonym(s):

Anion Exchange Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,180.00

$1,180.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$1,180.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$1,180.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Product Name

Hamilton® HPLC anion exchange column, PRP-X100, 10 μm, I.D. × L 4.1 mm × 250 mm

material

PSDVB/Trimethylammonium packing

manufacturer/tradename

Hamilton

packaging

1 ea of

parameter

5000 psi pressure

technique(s)

HPLC: suitable

I.D. × L

4.1 mm × 250 mm

matrix

polymeric

particle size

10 μm

pore size

100 Å

pH

1-13

separation technique

anion exchange

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service