Key Documents
HAM62576-01
Foot switch for Hamilton® Microlab® instruments
pack of 1 ea
Select a Size
1 EA
$333.00
$333.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$333.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
$333.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pack of 1 ea
cable L
1800 mm
General description
The Foot Switch is designed to work with Hamilton Microlab instruments. It is for applications where the user′s hands need to be free such as during an animal dosing experiment. The foot switch connects into the probe receptacle and sits on the ground. A simple press of the foot switch communicates with the pump and advances the programmed method.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)