Skip to Content
Merck
All Photos(2)

Documents

H3780

Sigma-Aldrich

2′-Hydroxyflavanone

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O3
CAS Number:
Molecular Weight:
240.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

SMILES string

Oc1ccccc1C2CC(=O)c3ccccc3O2

InChI

1S/C15H12O3/c16-12-7-3-1-5-10(12)15-9-13(17)11-6-2-4-8-14(11)18-15/h1-8,15-16H,9H2

InChI key

KZKWCKFDCPVDFJ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiyan Zhang et al.
International journal of clinical and experimental medicine, 8(9), 16955-16959 (2015-12-03)
Gastric carcinoma (GC) is the most common primary malignancy of the digestive tract, with increasing incidence in many countries. 3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay was used to assess inhibition of HepG2 cell proliferation by 2'-hydroxyflavanone. The STAT3 pathway was performed. 2'-hydroxyflavanone
Yusuke Iimura et al.
Journal of colloid and interface science, 508, 469-475 (2017-09-02)
The chiral selectivity of β-cyclodextrin (β-CD) in the water/hexane (3.0% THF) two-phase system is studied with chromatography and interfacial tension measurements. Chromatography using silica gel impregnated with aqueous β-CDas the stationary phase reveals that the chiral selectivity of this system
Tomohide Uno et al.
Biopharmaceutics & drug disposition, 36(8), 552-563 (2015-07-30)
CYP2A6 is a major hepatic member of the cytochrome P450 family in humans. Much variation in CYP2A6 levels and activity can be attributed to genetic polymorphisms of this gene. CYP2A6*25 comprises an amino acid substitution, F118L. To clarify the effect
Himangshu Sonowal et al.
Toxicology in vitro : an international journal published in association with BIBRA, 69, 104966-104966 (2020-08-18)
2'-Hydroxyflavanone (2-HF) is a natural flavonoid isolated from citrus fruits. Multiple studies have demonstrated that 2-HF with its anti-proliferative and pro-apoptotic effects prevent the growth of various cancers. Although 2-HF is a well known anti-oxidative and chemopreventive agent, its role
Mariana Silva et al.
Journal of chromatography. A, 1490, 166-176 (2017-02-17)
In this paper a chiral stationary phase (CSP) was prepared by the immobilization of a β-CD derivative (3,5-dimethylphenylcarbamoylated β-CD) onto the surface of amino-functionalized spherical ordered mesoporous silica (denoted as SM) via a urea linkage using the Staudinger reaction. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service