Skip to Content
Merck
All Photos(3)

Key Documents

H17309

Sigma-Aldrich

Hydrindantin

≥97%

Synonym(s):

2,2′-Dihydroxy-2,2′-biindan-1,1′,3,3′-tetrone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$142.00
25 G
$610.00

$142.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$142.00
25 G
$610.00

About This Item

Empirical Formula (Hill Notation):
C18H10O6
CAS Number:
Molecular Weight:
322.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$142.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97%

mp

252 °C (dec.) (lit.)

SMILES string

OC1(C(=O)c2ccccc2C1=O)C3(O)C(=O)c4ccccc4C3=O

InChI

1S/C18H10O6/c19-13-9-5-1-2-6-10(9)14(20)17(13,23)18(24)15(21)11-7-3-4-8-12(11)16(18)22/h1-8,23-24H

InChI key

LWFPYLZOVOCBPZ-UHFFFAOYSA-N

Application

Used in the determination of amino acids.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Giuseppina Sandri et al.
Polymers, 11(7) (2019-07-25)
Cutaneous wounds represent a major issue in medical care, with approximately 300 million chronic and 100 million traumatic wound patients worldwide, and microbial infections slow the healing process. The aim of this work was to develop electrospun scaffolds loaded with
Md Hasan Turabee et al.
Carbohydrate polymers, 203, 302-309 (2018-10-16)
The malignant gliomas are most destructive brain tumor having low drug response. The thermosensitive hydrogel from pluronic F127 (PF127) and N,N,N-trimethyl chitosan (TMC) is developed as a drug delivery system for anticancer drug docetaxel (DTX) to the glioblastoma multiforme. The
Shouchuan Li et al.
Soft matter, 15(2), 303-311 (2018-12-18)
Nano-clays (NCs) as a representative type of nano-materials are a source of inspiration for design of new biomedical materials with excellent performances. Research has shown that guanidinium ions (Gu+) can form non-covalent salt-bridge interactions with NCs, serving as "molecular glue"
Yihua Pei et al.
Journal of controlled release : official journal of the Controlled Release Society, 267, 133-143 (2017-08-12)
Methicillin-resistant Staphylococcus aureus (MRSA) infection is a serious threat to the public health. MRSA is particularly difficult to treat when it invades host cells and survive inside the cells. Although vancomycin is active against MRSA, it does not effectively kill
Angela Faccendini et al.
Pharmaceutics, 12(4) (2020-04-09)
Infections in nonhealing wounds remain one of the major challenges. Recently, nanomedicine approach seems a valid option to overcome the antibiotic resistance mechanisms. The aim of this study was the development of three types of polysaccharide-based scaffolds (chitosan-based (CH), chitosan/chondroitin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service