Skip to Content
Merck
All Photos(1)

Key Documents

H1256

Sigma-Aldrich

3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester

~90%

Synonym(s):

N-Succinimidyl 3-(4-hydroxyphenyl)propionate, N-[(4-Hydroxyhydrocinnamoyl)oxy]succinimide, Bolton-Hunter reagent precursor, Rudinger reagent, SHPP

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$260.00

$260.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$260.00

About This Item

Empirical Formula (Hill Notation):
C13H13NO5
CAS Number:
Molecular Weight:
263.25
Beilstein:
1545011
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$260.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

~90%

reaction suitability

reagent type: linker

mp

132-133 °C (lit.)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

Oc1ccc(CCC(=O)ON2C(=O)CCC2=O)cc1

InChI

1S/C13H13NO5/c15-10-4-1-9(2-5-10)3-8-13(18)19-14-11(16)6-7-12(14)17/h1-2,4-5,15H,3,6-8H2

InChI key

KYRUKRFVOACELK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester can be used to prepare 125I-labeled reagents, which are used in binding assay studies of proteins and peptides.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radioimmunoassay of iodinated gentamicin
Broughton, Alan and Strong, James E
Clinica Chimica Acta; International Journal of Clinical Chemistry, 66(1), 125-129 (1976)
E Marshall et al.
Molecular and biochemical parasitology, 15(3), 295-304 (1985-06-01)
The specificity of a range of 125I labelling techniques (Chloramine T, Iodogen, Bolton and Hunter reagent, lactoperoxidase and iodosulfanilic acid) to the surface of the filarial nematode Brugia pahangi was evaluated by autoradiography of sections of labelled worms and of
P Portolés et al.
Journal of immunological methods, 129(1), 105-109 (1990-05-08)
A method for the vectorial radioiodination of CD4 and other membrane proteins having few or no tyrosine residues in the extracytoplasmic domains is described. Incubation of the cells with sulfosuccinimidyl (hydroxyphenyl) propionate (sulfo-SHPP), a water-soluble derivative of the Bolton-Hunter reagent
M Rack et al.
Pflugers Archiv : European journal of physiology, 400(4), 403-408 (1984-04-01)
Several reagents that react with sulfhydryl and amino groups were applied to voltage-clamped single nerve fibres of the frog. The fibres were exposed to comparable amounts of the chemical reagents for relatively short times. 3-(p-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester (HPPS), a
Comparative methods for the radiolabeling of peptides.
A E Bolton
Methods in enzymology, 124, 18-29 (1986-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service