Skip to Content
Merck
All Photos(2)

Key Documents

GF98724730

Zinc

wire reel, 1m, diameter 0.125mm, as drawn, 99%

Synonym(s):

Zinc, ZN005100

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$337.00

$337.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$337.00

About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

$337.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

vapor pressure

1 mmHg ( 487 °C)

Assay

≥99%

form

wire

manufacturer/tradename

Goodfellow 987-247-30

resistivity

5.8 μΩ-cm, 20°C

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Granell
The Journal of sports medicine and physical fitness, 54(2), 232-237 (2014-02-11)
The role of zinc and copper has been shown essential in the scope of physical exercise. The outcomes of the studies about changes in the concentrations of these elements in blood, in physical effort situations, are sometimes discordant and seem
Douglas J Lanska et al.
Neurology, 82(13), 1175-1179 (2014-04-02)
Zinc-induced myeloneuropathy was recently (re)discovered and its pathophysiology elaborated as resulting from secondary copper deficiency. However, myelopathy was a recognized problem among European zinc-smelter workers in the late 19th century, although these early reports have been overlooked in recent studies
M Munirul Islam et al.
The Journal of nutrition, 144(5), 771-776 (2014-03-07)
Information is needed on breast milk zinc concentration and amount of zinc transferred to the infant in relation to the time since birth for both appropriate-for-gestational-age (AGA) and small-for-gestational-age (SGA) infants. Breast milk zinc concentration and total milk volume were
Shinya Tashiro et al.
Biochemistry, 53(14), 2218-2220 (2014-04-05)
Mutations of DJ-1 cause familial Parkinson's disease (PD), although the role of DJ-1 in PD remains unresolved. Very recent reports have shown that DJ-1 interacts with copper ions. This evidence opens new avenues to understanding the function of DJ-1 and
Priscilla K Walker et al.
Health physics, 106(4), 523-527 (2014-02-25)
Beam shaping in scattered and uniform scanned proton beam therapy (PBT) is made commonly by brass apertures. Due to proton interactions, these devices become radioactive and could pose safety issues and radiation hazards. Nearly 2,000 patient-specific devices per year are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service