The coil width is 200 mm, and for this product number, it would be 2 meters in length.
GF94703283
Manganin® - resistance alloy, Cu86Mn12Ni2
foil, 2m coil, 0.04mm thickness, annealed
Synonym(s):
CU060240
Select a Size
$2,100.00
Select a Size
About This Item
$2,100.00
Recommended Products
form
annealed
foil (coil)
manufacturer/tradename
Goodfellow 947-032-83
size × thickness
2 m × 0.04 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the width of the GF94703282 coil?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service