Skip to Content
Merck
All Photos(2)

Key Documents

GF93861850

Tantalum

foil, 10mm disks, thickness 0.125mm, annealed, 99.9%

Synonym(s):

Tantalum, TA000424, Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

≥99.9%

form

foil

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 938-618-50

resistivity

13.5 μΩ-cm, 20°C

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Observations on the use of tantalum foil in peripheral nerve surgery.
N C NORCROSS et al.
Journal of neurosurgery, 4(1), 69-71 (1947-01-01)
Hongbin Yu et al.
PloS one, 8(6), e66447-e66447 (2013-06-27)
We report a simple method to fabricate nano-porous tantalum oxide films via anodization with Ta foils as the anode at room temperature. A mixture of ethylene glycol, phosphoric acid, NH4F and H2O was used as the electrolyte where the nano-porous
The use of tantalum foil as a mechanical block for the separation of spinal cord and brain in chick embryos.
V HAMBURGER
The Anatomical record, 94, 355-355 (1946-03-01)
Xianfeng Yang et al.
Journal of the American Chemical Society, 131(34), 12048-12049 (2009-08-27)
A rutile microstructure with a novel cross-medal morphology was prepared via a hydrothermal method by using a Tantalum foil as both the doping source and the substrate. This rutile cross-medal prefers to grow along an unprecedented preferential orientation of <110>
Tantalum foil cuffs in peripheral nerve surgery.
E E CLIFFTON
Surgery, 23(3), 507-514 (1948-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service