Skip to Content
Merck
All Photos(2)

Key Documents

GF92772737

Rhenium

wire reel, 0.1m, diameter 0.125mm, annealed, 99.97%

Synonym(s):

Rhenium, RE005108

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$364.00

$364.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$364.00

About This Item

Empirical Formula (Hill Notation):
Re
CAS Number:
Molecular Weight:
186.21
MDL number:
UNSPSC Code:
12141737
PubChem Substance ID:
NACRES:
NA.23

$364.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

≥99.97%

form

wire

manufacturer/tradename

Goodfellow 927-727-37

bp

5596 °C (lit.)
5627 °C (lit.)

mp

3180 °C (lit.)

density

21.02 g/cm3 (lit.)

SMILES string

[ReH]

InChI

1S/Re

InChI key

WUAPFZMCVAUBPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malcolm Frier
Mini reviews in medicinal chemistry, 4(1), 61-68 (2004-02-03)
The favourable nuclear properties of copper-67 and rhenium-188 for therapeutic application are described, together with methods for the chemical synthesis of a number of derivatives. Survival from invasive bladder cancer has changed little over the past 20 years. The intravesicular
R Klett et al.
Rheumatology (Oxford, England), 46(10), 1531-1537 (2007-06-29)
Hypertrophy and inflammation of the synovium with various underlying pathologies - such as rheumatoid arthritis, osteoarthritis, haemophilia and spondyloarthropathy - can be treated successfully by radiosynoviorthesis (RSO). For medium-sized joints (shoulder, elbow, wrist, hip and ankle), the radionuclide of choice
Radiolabeling of monoclonal antibodies and fragments with technetium and rhenium.
G L Griffiths et al.
Bioconjugate chemistry, 3(2), 91-99 (1992-03-01)
Mark Bartholomä et al.
Chemical communications (Cambridge, England), (5)(5), 493-512 (2009-03-14)
Radiolabeled biomolecules can be used to visualize a variety of diseases through interaction with specific cell receptors. A key step is the introduction of a molecular entity that allows facile labeling with the medically useful radionuclide (99m)Tc without significant alteration
N Iznaga-Escobar
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 54(3), 399-406 (2001-02-24)
188Re is one of the radioisotopes expected to emerge as useful for therapy. Development of new radiopharmaceuticals based on 188Re depends on the radiolabeling methods used, which would give stable complexes having predefined radiochemical properties and in vitro and in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service