Skip to Content
Merck
All Photos(2)

Key Documents

GF91382069

Cobalt

wire reel, 0.1m, diameter 0.1mm, hard, 99.99+%

Synonym(s):

Cobalt, CO005085

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$427.00

$427.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$427.00

About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
MDL number:
UNSPSC Code:
12141710
PubChem Substance ID:
NACRES:
NA.23

$427.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

≥99.99%

form

wire

manufacturer/tradename

Goodfellow 913-820-69

resistivity

6.24 μΩ-cm, 20°C

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao-Di Wu et al.
Journal of nanoscience and nanotechnology, 14(6), 4097-4100 (2014-04-18)
Cocrystal nanofibres of cobalt octaethylporphyrin and tetracyanoquinodimethane were prepared by a facile solution method and fully characterized by SEM, AFM, XRD, Raman, EDX, and UV-vis-NIR. The as-prepared cocrystal nanofibres had smooth surfaces and uniform dimension. When incorporated into prototype devices
Anthony F Pizon et al.
Journal of medical toxicology : official journal of the American College of Medical Toxicology, 9(4), 416-417 (2013-11-22)
Prosthetic hip-associated cobalt toxicity (PHACT) is gaining recognition due to the use of metal-on-metal total hip replacements. Identifying true toxicity from merely elevated cobalt levels can be extremely difficult due to the lack of available data. An extensive review of
Kazuhiro Yamada
Metal ions in life sciences, 13, 295-320 (2014-01-29)
The primarily function of cobalt in humans is based on its role in cobalamin (Cbl, vitamin B12). Therefore, this chapter will focus on the physiological roles of Cbl and the importance of cobalt in human health. Cbl acts as the
Kenneth M Unice et al.
Chemico-biological interactions, 216, 53-74 (2014-04-15)
An updated biokinetic model for human exposures to cobalt (Co) was developed based on a comprehensive set of human pharmacokinetics data collected from five male and five female volunteers who ingested ∼1 mg Co/day of a Co supplement for 3
John J Devlin et al.
Journal of medical toxicology : official journal of the American College of Medical Toxicology, 9(4), 405-415 (2013-11-14)
Safety concerns regarding cobalt-containing metal alloy hip prosthetics (Co-HP) have resulted in product recalls, a medical device alert, and issuance of guidance for clinicians. Recently, cases of suspected prosthetic hip-associated cobalt toxicity (PHACT) from Co-HP have been reported. Although little

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service