Skip to Content
Merck
All Photos(2)

Key Documents

GF80118828

Carbon

rod, 150mm, diameter 10.0mm, graphite, 100%

Synonym(s):

Activated charcoal, C 007930, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

100%

form

rod

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 801-188-28

resistivity

1375 μΩ-cm, 20°C (graphite)

size diam.

150 mm × 10.0 mm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi-Jung Su et al.
Journal of nanoscience and nanotechnology, 14(6), 4201-4206 (2014-04-18)
Carbon nanofibers (CNFs) have wide applications in energy storage devices, electrically conducting composites, selective adsorbents, and catalyst supports. Catalytic chemical vapor deposition was carried out in this work to synthesize CNFs at mild temperatures of 700 and 800 degrees C.
Ivar Zekker et al.
Environmental technology, 35(9-12), 1565-1576 (2014-04-08)
Maintaining stability of low concentration (< 1 g L(-1)) floccular biomass in the nitritation-anaerobic ammonium oxidation (anammox) process in the sequencing batch reactor (SBR) system for the treatment of high COD (> 15,000 mg O2 L(-1)) to N (1680 mg
Kimberly T Barrett et al.
Nature, 509(7498), 71-75 (2014-04-22)
One of the most well-recognized stereogenic elements in a chiral molecule is an sp(3)-hybridized carbon atom that is connected to four different substituents. Axes of chirality can also exist about bonds with hindered barriers of rotation; molecules containing such axes
Gregory R Wagner et al.
Molecular cell, 54(1), 5-16 (2014-04-15)
Cellular proteins are decorated with a wide range of acetyl and other acyl modifications. Many studies have demonstrated regulation of site-specific acetylation by acetyltransferases and deacetylases. Acylation is emerging as a new type of lysine modification, but less is known
Deivasigamani Ranjith Kumar et al.
Journal of nanoscience and nanotechnology, 14(7), 5059-5069 (2014-04-25)
Iron oxide (Fe3O4) nanoparticles are prepared by nucleation method using oleylamine (OLA) as the capping agent. The OLA capped Fe3O4 nanoparticles (OLA-Fe3O4) are characterized by Fourier Transform Infrared (FT-IR) spectroscopy, X-ray Diffraction (XRD) and High Resolution Transmission Electron Microscopic (HRTEM)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service