GF79419695
Tin
foil, 25mm disks, thickness 1.0mm, as rolled, 99.99+%
Synonym(s):
Tin, SN000440
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
PubChem Substance ID:
NACRES:
NA.23
Assay
99.99%
form
foil
manufacturer/tradename
Goodfellow 794-196-95
resistivity
11 μΩ-cm, 20°C
diam. × thickness
25 mm × 1.0 mm
bp
2270 °C (lit.)
mp
231.9 °C (lit.)
density
7.310 g/mL at 25 °C (lit.)
SMILES string
[Sn]
InChI
1S/Sn
InChI key
ATJFFYVFTNAWJD-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mercury contamination removed with tin foil.
R Grajower et al.
Operative dentistry, 9(3), 101-104 (1984-01-01)
Andrey A Gurtovenko et al.
The Journal of chemical physics, 130(21), 215107-215107 (2009-06-11)
The electrostatic properties of lipid membranes are of profound importance as they are directly associated with membrane potential and, consequently, with numerous membrane-mediated biological phenomena. Here we address a number of methodological issues related to the computation of the electrostatic
Jianan Liu et al.
Animal reproduction science, 134(3-4), 197-202 (2012-08-28)
Cryopreservation of ovarian tissue has been the only effective way of ex situ conservation of female germplasm in avian species. A novel needle-in-straw (NIS) vitrification method was developed to store tissue in straws instead of cryovials. Fragments of ovarian tissue
J Strang et al.
Addiction (Abingdon, England), 92(6), 673-683 (1997-06-01)
The history of heroin smoking and the subsequent development and spread of 'chasing the dragon' are examined. The first heroin smoking originated in Shanghai in the 1920s and involved use of porcelain bowls and bamboo tubes, thereafter spreading across much
M R Krigman et al.
Neurotoxicology, 5(2), 129-139 (1984-01-01)
The toxicology of tin is almost entirely the toxicology of the organic compounds of tin, for the metal itself and its inorganic compounds appear to be nearly harmless for practical purposes. Furthermore, the neurotoxicity of organotin is essentially that of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service