Skip to Content
Merck
All Photos(2)

Key Documents

GF77697680

Germanium

microfoil, 25x25mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%

Synonym(s):

Germanium, GE004600

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ge
CAS Number:
Molecular Weight:
72.64
MDL number:
UNSPSC Code:
12141716
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

100%

form

foil

manufacturer/tradename

Goodfellow 776-976-80

resistivity

53 Ω-cm, 20°C

size × thickness

25 x 25 mm × 0.25 mm

bp

2830 °C (lit.)

mp

937 °C (lit.)

density

5.35 g/mL at 25 °C (lit.)

SMILES string

[Ge]

InChI

1S/Ge

InChI key

GNPVGFCGXDBREM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunfei Jiang et al.
Plant, cell & environment, 38(11), 2387-2397 (2015-06-18)
Pea (Pisum sativum L.) is a major legume crop grown in a semi-arid climate in Western Canada, where heat stress affects pollination, seed set and yield. Seed set and pod growth characteristics, along with in vitro percentage pollen germination, pollen
Liqun Zhou et al.
Journal of dentistry, 43(8), 973-980 (2015-06-10)
The present study examined (1) the chemical interaction between three self-etching adhesives and sodium hypochlorite (NaOCl)-deproteinised dentine, and (2) the influence of NaOCl treatment on bond strength of self-etching adhesives with/without adhesive functional monomers to dentine. Caries-free dentine disks (control)
S H Tao et al.
Regulatory toxicology and pharmacology : RTP, 25(3), 211-219 (1997-06-01)
Germanium-containing dietary supplements became popular in the 1970s in Japan and later in other countries, as elixirs for certain diseases (e.g., cancer and AIDS). Germanium is not an essential element. Its acute toxicity is low. However, at least 31 reported
Gui-Di Yang et al.
Ecotoxicology and environmental safety, 120, 7-12 (2015-05-31)
Arsenite [As (III)] oxidation can be accelerated by bacterial catalysis, but the effects of the accelerated oxidation on arsenic toxicity and translocation in rice plants are poorly understood. Herein we investigated how an arsenite-oxidizing bacterium, namely Brevibacillus laterosporus, influences As
Li Z Pei et al.
Recent patents on nanotechnology, 6(1), 44-59 (2011-10-26)
Ge nanowires exhibit wide application potential in the fields of nanoscale devices due to their excellently optical and electrical properties. This article reviews the recent progress and patents of Ge nanowires. The recent progress and patents for the synthesis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service