Skip to Content
Merck
All Photos(2)

Key Documents

GF71906542

Zinc

wire reel, 20m, diameter 2.0mm, extruded, 99.9%

Synonym(s):

Zinc, ZN005135

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$482.00

$482.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$482.00

About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

$482.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

vapor pressure

1 mmHg ( 487 °C)

Assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 719-065-42

resistivity

5.8 μΩ-cm, 20°C

L × diam.

20 m × 2.0 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johan Hattne et al.
Nature methods, 11(5), 545-548 (2014-03-19)
X-ray free-electron laser (XFEL) sources enable the use of crystallography to solve three-dimensional macromolecular structures under native conditions and without radiation damage. Results to date, however, have been limited by the challenge of deriving accurate Bragg intensities from a heterogeneous
Priscilla K Walker et al.
Health physics, 106(4), 523-527 (2014-02-25)
Beam shaping in scattered and uniform scanned proton beam therapy (PBT) is made commonly by brass apertures. Due to proton interactions, these devices become radioactive and could pose safety issues and radiation hazards. Nearly 2,000 patient-specific devices per year are
Kohei Takeshita et al.
Nature structural & molecular biology, 21(4), 352-357 (2014-03-04)
The voltage-gated proton channel Hv1 (or VSOP) has a voltage-sensor domain (VSD) with dual roles of voltage sensing and proton permeation. Its gating is sensitive to pH and Zn(2+). Here we present a crystal structure of mouse Hv1 in the
Yongmei Zhao et al.
Stroke, 45(4), 1139-1147 (2014-03-20)
Zinc has been reported to possess both neurotoxic and neuroprotective capabilities. The effects of elevated intracellular zinc accumulation following transient focal cerebral ischemia remain to be fully elucidated. Here, we investigated whether removing zinc with the membrane-permeable zinc chelator, N,N,N',N'-tetrakis(2-pyridylmethyl)ethylenediamine
Ronny Lehneck et al.
The FEBS journal, 281(7), 1759-1772 (2014-02-11)
Carbonic anhydrases (CAs) are metalloenzymes catalyzing the reversible hydration of carbon dioxide to bicarbonate (hydrogen carbonate) and protons. CAs have been identified in archaea, bacteria and eukaryotes and can be classified into five groups (α, β, γ, δ, ζ) that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service