Skip to Content
Merck
All Photos(1)

Key Documents

GF68527187

Alumina

powder, 0.1 max. part. size (micron), weight 50 g, condition 99.99% gamma alumina powder.

Synonym(s):

AL606022

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$255.00

$255.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$255.00

About This Item

UNSPSC Code:
11181511
NACRES:
NA.23

$255.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99.99%

form

powder

manufacturer/tradename

Goodfellow 685-271-87

weight

50 g

max. part. size

0.1 (micron)

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF74339488GF62302531GF41348864
form

powder

form

powder

form

powder

form

powder

assay

≥99.99%

assay

99.9%

assay

≥99.99%

assay

≥99.99%

manufacturer/tradename

Goodfellow 685-271-87

manufacturer/tradename

Goodfellow 743-394-88

manufacturer/tradename

Goodfellow 623-025-31

manufacturer/tradename

Goodfellow 413-488-64

weight

50 g

weight

50 g

weight

20 g

weight

200 g

max. part. size

0.1 (micron)

max. part. size

-

max. part. size

0.1 (micron)

max. part. size

0.1 μm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service