Skip to Content
Merck
All Photos(2)

Key Documents

GF65129104

Tungsten

washer, length 25 pcs, size m3, outside diameter 6.0mm

Synonym(s):

W 00WA03, W

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$467.00

$467.00


Estimated to ship onMarch 24, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$467.00

About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
MDL number:
UNSPSC Code:
31161807
PubChem Substance ID:
NACRES:
NA.23

$467.00


Estimated to ship onMarch 24, 2025


Request a Bulk Order

packaging

pkg of 25

manufacturer/tradename

Goodfellow 651-291-04

resistivity

4.9 μΩ-cm, 20°C

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[W]

InChI

1S/W

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service