Skip to Content
Merck
All Photos(2)

Key Documents

GF62383417

Iron

powder, 6-8 μm mean particle size, weight 2000 g, purity 99.0+%

Synonym(s):

FE006041

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,280.00

$1,280.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,280.00

About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

$1,280.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99.0%

form

powder

manufacturer/tradename

Goodfellow 623-834-17

resistivity

9.71 μΩ-cm

weight

2000 g

mean particle size

6-8 μm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service