GF58769443
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3
foil, 10m coil, 12mm coil width, 0.2mm thickness, annealed
Synonym(s):
FF210321
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
annealed
foil (coil)
manufacturer/tradename
Goodfellow 587-694-43
L × W × thickness
10 m × 12 mm × 0.2 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service