GF58757675
Magnesium
foil, thickness 0.003 mm, 15 mm diameter, purity 99.9%
Synonym(s):
MG000100
About This Item
vapor pressure
1 mmHg ( 621 °C)
Assay
99.9%
form
foil
autoignition temp.
950 °F
manufacturer/tradename
Goodfellow 587-576-75
resistivity
4.46 μΩ-cm, 20°C
diam.
15 mm
thickness
0.003 mm
diameter
15 mm
bp
1090 °C (lit.)
mp
648 °C (lit.)
density
1.74 g/mL at 25 °C (lit.)
SMILES string
[Mg]
InChI
1S/Mg
InChI key
FYYHWMGAXLPEAU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service