GF51231833
Molybdenum
wire reel, 50m, diameter 0.025mm, gold plated, 99.95%
Synonym(s):
Molybdenum, MO005220
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.95%
form
wire
manufacturer/tradename
Goodfellow 512-318-33
resistivity
5.0 μΩ-cm, 20°C
L × diam.
50 m × 0.025 mm
bp
4612 °C (lit.)
mp
2617 °C (lit.)
density
10.3 g/mL at 25 °C (lit.)
SMILES string
[Mo]
InChI
1S/Mo
InChI key
ZOKXTWBITQBERF-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mohammad-Reza Rashidi et al.
Expert opinion on drug metabolism & toxicology, 6(2), 133-152 (2010-01-26)
Molybdenum hydroxylases, aldehyde oxidase and xanthine oxidase, are metalloflavoproteins that catalyze both oxidation and reduction of a broad range of drugs and other xenobiotics indicating the importance of these enzymes in drug oxidation, detoxification and activation. Both enzymes are also
Ralf R Mendel
The Journal of biological chemistry, 288(19), 13165-13172 (2013-03-30)
The transition element molybdenum needs to be complexed by a special cofactor to gain catalytic activity. Molybdenum is bound to a unique pterin, thus forming the molybdenum cofactor (Moco), which, in different variants, is the active compound at the catalytic
Chantal Iobbi-Nivol et al.
Biochimica et biophysica acta, 1827(8-9), 1086-1101 (2012-12-04)
Molybdenum cofactor (Moco) biosynthesis is an ancient, ubiquitous, and highly conserved pathway leading to the biochemical activation of molybdenum. Moco is the essential component of a group of redox enzymes, which are diverse in terms of their phylogenetic distribution and
Viswanathan S Saji et al.
ChemSusChem, 5(7), 1146-1161 (2012-06-14)
The electrochemical behaviors of molybdenum and its oxides, both in bulk and thin film dimensions, are critical because of their widespread applications in steels, electrocatalysts, electrochromic materials, batteries, sensors, and solar cells. An important area of current interest is electrodeposited
Maria Lyra et al.
Hellenic journal of nuclear medicine, 14(1), 49-55 (2011-04-23)
The sleeping giant of molybdenum-99 ((99)Mo) production is grinding to a halt and the world is wondering how this happened. Fewer than 10 reactors in the world are capable of producing radio nuclides for medicine; approximately 50% of the world's
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service