GF43826542
Bismuth
foil, thickness 0.009 mm, size 50 × 50 mm, purity 99.97%
Synonym(s):
BI000110
Select a Size
Select a Size
About This Item
vapor pressure
<0.1 mmHg ( 20 °C)
Assay
99.97%
form
foil
manufacturer/tradename
Goodfellow 438-265-42
resistivity
129 μΩ-cm, 20°C
size
50 × 50 mm
thickness
0.009 mm
bp
1560 °C (lit.)
mp
271 °C (lit.)
density
9.8 g/mL at 25 °C (lit.)
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | GF35950507 | GF93812687 | GF93887601 |
---|---|---|---|
50 mm | 200 mm | 500 mm | 100 mm |
Goodfellow 439-511-03 | Goodfellow 359-505-07 | Goodfellow 938-126-87 | Goodfellow 938-876-01 |
rod | rod | rod | rod |
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service