Skip to Content
Merck
All Photos(1)

Documents

GF42328986

Monel® alloy 400, Ni65Cu33Fe2

200mm tube, 7.1mm outside diameter, 0.27mm wall thickness, 6.56mm inside diameter, hard

Synonym(s):

NI117620

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 423-289-86

L

200 mm

O.D. × I.D. × wall thickness

7.1 mm × 6.5 mm × 0.27 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Monel is a registered trademark of Special Metals Corporation
Monel is a registered trademark of Huntington Alloys Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service