GF41005538
Zirconium
rod, 200mm, diameter 9.5mm, centerless ground, 99.2%
Synonym(s):
Zirconium, ZR007930
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
99.2%
form
rod
manufacturer/tradename
Goodfellow 410-055-38
resistivity
40 μΩ-cm, 20°C
L × diam.
200 mm × 9.5 mm
bp
4377 °C (lit.)
mp
1852 °C (lit.)
density
6.5 g/mL at 25 °C (lit.)
SMILES string
[Zr]
InChI
1S/Zr
InChI key
QCWXUUIWCKQGHC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Glypican-3-targeted 89Zr PET imaging of hepatocellular carcinoma: where antibody imaging dares to tread.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(5), 708-709 (2014-03-26)
Indian journal of dental research : official publication of Indian Society for Dental Research, 22(1), 140-143 (2011-04-29)
The demand for esthetic restorations has resulted in an increased use of dental ceramics for anterior and posterior restorations. A few decades ago, all-ceramic restorations were restricted to treatment in the anterior region, but now all-ceramic restorations can be made
Medicina oral, patologia oral y cirugia bucal, 17(2), e246-e250 (2011-12-07)
The increase of aesthetic demands, together with the successful outcome of current implants, has renewed interest in the search for new materials with enough mechanical properties and better aesthetic qualities than the materials customarily used in implanto-prosthetic rehabilitation. Among these
Current radiopharmaceuticals, 4(2), 131-139 (2011-12-24)
Positron emission tomography (PET) imaging with radiolabeled monoclonal antibodies has always been a dynamic area in molecular imaging. With decay half-life (3.3 d) well matched to the circulation half-lives of antibodies (usually on the order of days), (89)Zr has been
Journal of dental research, 93(4), 329-334 (2014-02-25)
Dental zirconia can no longer be considered un-bondable to tooth tissue. In literature, an increasing number of papers indeed report on the bonding effectiveness of different luting techniques to zirconia. We aimed to disclose general trends in bonding to zirconia
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service