Skip to Content
Merck

Skip To

GF39991663

Nickel

crucible, cylindrical, length 1 pcs, outside diameter 21mm

Synonym(s):

NI008605

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
$230.00

About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
11101706
MDL number:
eCl@ss:
38190701

$230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1

manufacturer/tradename

Goodfellow 399-916-63

resistivity

6.97 μΩ-cm, 20°C

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF21869130GF75117427GF04598859
manufacturer/tradename

Goodfellow 399-916-63

manufacturer/tradename

Goodfellow 218-691-30

manufacturer/tradename

Goodfellow 751-174-27

manufacturer/tradename

Goodfellow 045-988-59

density

8.9 g/mL at 25 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

resistivity

6.97 μΩ-cm, 20°C

resistivity

6.97 μΩ-cm, 20°C

resistivity

6.97 μΩ-cm, 20°C

resistivity

6.97 μΩ-cm, 20°C

mp

1453 °C (lit.)

mp

1453 °C (lit.)

mp

1453 °C (lit.)

mp

1453 °C (lit.)

bp

2732 °C (lit.)

bp

2732 °C (lit.)

bp

2732 °C (lit.)

bp

2732 °C (lit.)

packaging

pkg of 1

packaging

pkg of 1

packaging

pkg of 1

packaging

pkg of 1


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters