GF39917818
Copper-nickel alloy, Cu70Ni30
1000mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, hard
Synonym(s):
CU137200
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352300
NACRES:
NA.23
Recommended Products
form
tubes
manufacturer/tradename
Goodfellow 399-178-18
L
1000 mm
O.D. × I.D. × thickness
1.0 mm × 0.8 mm × 0.1 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service