Skip to Content
Merck
All Photos(2)

Documents

GF38685134

Zirconium

foil, 6mm disks, thickness 0.02mm, 99.2%

Synonym(s):

Zirconium, ZR000200

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

Assay

99.2%

form

foil

manufacturer/tradename

Goodfellow 386-851-34

resistivity

40 μΩ-cm, 20°C

diam. × thickness

6 mm × 0.02 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glypican-3-targeted 89Zr PET imaging of hepatocellular carcinoma: where antibody imaging dares to tread.
Bernadette V Marquez et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(5), 708-709 (2014-03-26)
R Velázquez-Cayón et al.
Medicina oral, patologia oral y cirugia bucal, 17(2), e246-e250 (2011-12-07)
The increase of aesthetic demands, together with the successful outcome of current implants, has renewed interest in the search for new materials with enough mechanical properties and better aesthetic qualities than the materials customarily used in implanto-prosthetic rehabilitation. Among these
Anjana Raut et al.
Indian journal of dental research : official publication of Indian Society for Dental Research, 22(1), 140-143 (2011-04-29)
The demand for esthetic restorations has resulted in an increased use of dental ceramics for anterior and posterior restorations. A few decades ago, all-ceramic restorations were restricted to treatment in the anterior region, but now all-ceramic restorations can be made
D B Zhu et al.
Journal of nanoscience and nanotechnology, 14(5), 3703-3706 (2014-04-17)
Functionalized ceramic dental crown was successfully fabricated through selective slurry extrusion (SSE) based technique of solid freeform fabrication (also known as rapid prototyping). After sintering, the decomposed tourmaline powders were embedded in ZrO2 matrix. The far infrared emission properties of
M Inokoshi et al.
Journal of dental research, 93(4), 329-334 (2014-02-25)
Dental zirconia can no longer be considered un-bondable to tooth tissue. In literature, an increasing number of papers indeed report on the bonding effectiveness of different luting techniques to zirconia. We aimed to disclose general trends in bonding to zirconia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service