Skip to Content
Merck
All Photos(1)

Key Documents

GF36350175

Stainless Steel - 17-7PH alloy, FeCr17Ni7

foil, 20m coil, 25mm coil width, 0.15mm thickness, as rolled - condition c, precipitation hardenable

Synonym(s):

FE280321

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

description

as rolled - condition c (precipitation hardenable)

form

foil (coil)

manufacturer/tradename

Goodfellow 363-501-75

L × W × thickness

20 m × 25 mm × 0.15 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service