Skip to Content
Merck
All Photos(1)

Key Documents

GF30045855

Brass alloy, Cu70Zn30

sphere, 3.0mm diameter, 5.08μm sphericity, precision sphere, grade 200, 500pcs

Synonym(s):

CU086840

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

form

spheres (grade 200)

quality

precision sphere

packaging

1 ea of 500 pieces

manufacturer/tradename

Goodfellow 300-458-55

diam.

3.0 mm

sphericity

5.08 μm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service